Learn More
Cole-Parmer™ Low-Density Polyethylene Bags
Use these bags for general storage of solids. Cole-Parmer&trade Low-Density Polyethylene Bags feature a strong bottom seal.
Brand: Cole-Parmer™ 06505-06
Additional Details : Weight : 0.80000kg
Description
- Seal with portable heat sealer or bag ties
Specifications
LDPE Bag | |
Polyethylene | |
1000 Each | |
127 mm |
Clear | |
0.050 mm | |
127 x 102mm | |
102 mm |