missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius Stedim Biotech™ Polyethersulfone (PES) Membrane Filters for e.motion Dispenser
General purpose membrane that provides excellent performance with most solutions when retentate recovery is of primary importance.
Brand: Sartorius Stedim Biotech™ 15406Z-47----SCM
Additional Details : Weight : 0.14200kg
Description
- Suitable for e.motion dispensers
- General purpose membrane that provides excellent performance with most solutions when retentate recovery is of primary importance.
- PES membranes exhibit no hydrophobic or hydrophilic interactions and are usually preferred for their low fouling characteristics, exceptional flux and broad pH range.
- Suitable for biopharma, research, buffer exchange, protein/DNA concentration
Specifications
White | |
0.45 μm | |
Hydrophilic | |
47 mm |
PES | |
Sterile | |
300/pk. | |
47 mm |