Learn More
Buerkle™ Polyacetal Hose Clips
Shop All Bürkle Products
Click to view available options
Tubing Size:
10 to 11 mm
11 to 12 mm
12 to 14 mm
13 to 15 mm
15 to 17 mm
17 to 19 mm
18 to 20 mm
20 to 22 mm
23 to 25 mm
6 to 6.5 mm
9 to 10 mm
Unit Size:
Pack of 10
Description
- Hose clips made of Polyacetal (POM)
- Featured in varying diameters
- POM is white, sterilizable, does not rust, permanently elastic and hygienic.
Specifications
Specifications
| Material | Polyacetal |
| For Use With | Hoses |
| Packaging Quantity | 10 Pc./Pk. |
| Tubing Size | 13 to 15 mm |
| Color | White |
| Product Type | Hose Clip |