Learn More
JG Finneran Associates™ Poly Crimp™ Seals
Brand: JG Finneran Associates™ 5560-11
Additional Details : Weight : 0.50000kg
Specifications
PTFE/Silicone/PTFE | |
PTFE, Silicone | |
11 mm |
Clear | |
1000/Pk. | |
12 x 32 mm Sample Vials with Crimp Style Finish |