Learn More
Trajan Scientific and Medical™ Plunger replacement for Shimadzu autosampler syringe PTFE plunger tip 10μL SGE Diamond MS
SGE syringe replacement plungers for gas tight syringes
Brand:  Trajan Scientific and Medical™ 0355180
Additional Details : Weight : 0.05000kg
Product Code. 13137956
  • 62.30€ / Each
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match specific syringe barrel to maintain gas tight seal
  • For use with Shimadzu Autosampler Syringes
Specifications
Specifications
PTFE (Plunger)
10 μL
Gas Tight
Replacement Plunger
Shimadzu Autosampler Syringe