Learn More
Kartell™ Plastilab™ Untapered Y-Shaped Tubing Connectors
Product Code. 12543307
Change view
Click to view available options
Diameter (Metric):
4 mm
8 mm
10 mm
14 mm
Packaging Quantity:
1
10/Pk.
100/Pk.
Unit Size:
Pack of 10
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Packaging Quantity unitSize
12543307 14 mm 100/Pk. Pack of 100
12080582 10 mm 1 Pack of 100
11914349 8 mm 10/Pk. Pack of 10
11944359 4 mm 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12543307 Supplier Kartell™ Supplier No. 0052600
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
Designed for use with hose system and vacuum lines.
  • Made from polypropylene
  • Autoclavable
  • Nominal dimensions
Compliance
Meet Food and Agriculture regulations
Specifications
Certifications/Compliance Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD
Diameter (Metric) 14 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With (Application) Useful for any hoses system and vacuum line
Packaging Quantity 100/Pk.
Port Size 9.7mm bore
Shape Y
Product Type Untapered Y Tubing Connector