Learn More
Fisherbrand™ Nylon Sieve Filter, 1m x 1m
Product Code. 11795488 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Pore Size:
5 μm
11 μm
20 μm
25 μm
30 μm
41μm
60 μm
70 μm
80 μm
100 μm
150 μm
200 μm
250 μm
300 μm
400 μm
500 μm
1000 μm
1250 μm
1400 μm
1550 μm
1700 μm
1800 μm
2000 μm
Unit Size:
Each
23 product options available for selection
Product selection table with 23 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size Unit Size
11795488 100 μm Each
11785498 1000 μm Each
11705498 150 μm Each
11705488 11 μm Each
11715488 20 μm Each
11715498 200 μm Each
11755518 2000 μm Each
11725498 250 μm Each
11735488 30 μm Each
11745488 41μm Each
11795478 5 μm Each
11755498 500 μm Each
11765488 60 μm Each
11775488 70 μm Each
11705508 1400 μm Each
11745498 400 μm Each
11715508 1550 μm Each
11725488 25 μm Each
11725508 1700 μm Each
11735498 300 μm Each
11735508 1800 μm Each
11785488 80 μm Each
11795498 1250 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 23 options available.
23 options
Product Code. 11795488
  • ONLINE EXCLUSIVE PRICE
    211.01€
    99.65€ / Pack of 1
    Save 111.36€
    52% Off
In Stock
Add to basket
Edge
Add to basket
For sieving or precision filtration of biological and chemical solutions. Fisherbrand™ Nylon Sieve Filters, 1m x 1m are available in a comprehensive range of pore sizes.
  • Resistant to aggressive solvents
  • Excellent compatibility with alcohols
  • Operating temperature of -40° to +115°C
  • Applications include solvent filtration and particle analysis
Specifications
Pore Size 100 μm
Frame Material Plastic
Mesh Material Nylon