Learn More
Multiroir Controlec™ Plastic Mobile Storage Cart
Product Code. 16437570
Click to view available options
No. of Drawers:
3
Unit Size:
Each
Product Code. 16437570
Brand: Multiroir Controlec™ 18105
Edge
The Multiroir multi-drawer storage cart is ideal for general storage of items that can easily be seen through the translucent plastic drawers. The cart is on casters so it can easily be moved around the workspace.
  • Black housing on casters
  • Translucent drawers make objects easily visible
  • Lightweight
  • Resistant to shocks, cleaning products and temperature variations
  • Made in France
Specifications
Autoclavable Not Autoclavable
Capacity (Metric) 15L drawers
Color Black
Description Black tower with 3 transparent drawers
Dimensions (L x W x H) 40 x 29 x 67 cm (with casters)
For Use With (Application) General storage
Material Polypropylene
No. of Casters 4 casters, 360°
No. of Drawers 3
Weight (Metric) 2.4 kg