Learn More
Plas Labs™ Broome-Style Rodent Restrainers
Product Code. 12505846
Change view
Click to view available options
Diameter (English):
1 in.
2 in.
2.5 in.
Length (English):
3.75 in.
8 in.
8.5 in.
Weight Range:
125 to 250 g
15 to 30 g
250 to 500 g
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Length (English) Weight Range Unit Size
12505846 1 in. 3.75 in. 15 to 30 g Each
13204979 2 in. 8 in. 125 to 250 g Each
15193745 2.5 in. 8.5 in. 250 to 500 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12505846 Supplier Plas Labs™ Supplier No. 551BSRR
  • ONLINE EXCLUSIVE PRICE
    165.50€
    93.84€ / Pack of 1
    Save 71.66€
    43% Off
In Stock
Add to basket
Edge
Add to basket
Ideal for injections or blood sampling
  • Offer easy access to rodent
  • Prevent rodent from turning
  • Provide safety to the animal handler
  • Manufactured from clear materials to provide complete visibility of animal
Specifications
Color Clear
For Use With (Application) Injections or Blood Sampling
Type Rodent Restrainer
Material Noryl™ Plastic
Sterilization Method Chemically sterilized or washed at 180°F
Style Broome
Length (English) 3.75 in.
Length (Metric) 9.52 cm
Diameter (English) 1 in.
Diameter (Metric) 2.54 cm
Weight Range 15 to 30 g