Learn More
Hamilton™ 5000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Essential for precise pipetting. Hamilton™ 5000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
Brand:  Hamilton™ 184020
Additional Details : Weight : 0.01000kg
Product Code. 13484646
  • 168.00€ / Pack of 720
Estimated Shipment: 11-06-2024
Description
  • 118mm tip length
  • Color code label: green
  • Usable with 5000μL pipetting channels
  • Pressure-based Liquid Level Detection (pLLD) and capacitive (cLLD) with conductive (black) version
  • Process safety offered by Monitored Air Displacement and Total Aspiration and Dispense Monitoring
Specifications
Specifications
5000 μL
Non-sterile
Green
Non-Filtered