Learn More
Hamilton™ 5000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Essential for precise pipetting. Hamilton™ 5000μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
Brand: Hamilton™ 184020
Additional Details : Weight : 0.01000kg
Description
- 118mm tip length
- Color code label: green
- Usable with 5000μL pipetting channels
- Pressure-based Liquid Level Detection (pLLD) and capacitive (cLLD) with conductive (black) version
- Process safety offered by Monitored Air Displacement and Total Aspiration and Dispense Monitoring
Specifications
5000 μL | |
Non-sterile | |
Green |
Non-Filtered | |
Black | |
Single-use |