Learn More
Hamilton™ 50μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Essential for precise pipetting. Hamilton™ 50μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
Brand:  Hamilton™ 235989
Additional Details : Weight : 0.01000kg
Product Code. 15167416
  • 760.00€ / Pack of 7680
In Stock
Description
  • 39.5mm tip length
  • Color code label: light red
  • Usable with CO-RE 384 Probe Head
  • Fine, thin tip facilitates dosing of micro volumes
  • Capacitive LLD (cLLD) with conductive (black) version
  • Process safety offered by Monitored Air Displacement
Specifications
Specifications
50 μL
Non-sterile
Conductive
Single-use
Non-Filtered