Learn More
Hamilton™ 50μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips
Essential for precise pipetting. Hamilton™ 50μL Conductive Non-Filter, Non-Sterile CO-RE Pipet Tips provide enhanced pipetting safety and reliability with the CO-RE PerfectTouch system.
Brand: Hamilton™ 235989
Additional Details : Weight : 0.01000kg
Description
- 39.5mm tip length
- Color code label: light red
- Usable with CO-RE 384 Probe Head
- Fine, thin tip facilitates dosing of micro volumes
- Capacitive LLD (cLLD) with conductive (black) version
- Process safety offered by Monitored Air Displacement
Specifications
50 μL | |
Non-sterile | |
Conductive | |
Single-use |
Non-Filtered | |
Black | |
Red |