Learn More
Abnova™ PIK3CB/CEN3q (Texas Red/FITC) FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Brand:  Abnova™ FG0142.100uL
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 16013225
  • 1475.00€ / 100µL
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
Includes: DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe
Description
Description
FISH is a technique used to identify and localize the presence or absence of specific DNA sequences on cells and tissues. Abnova has developed a range of FISH probes for the detection of gene amplification, loss, and translocation. Each FISH probe has a pair of locus-specific, fluorophore-labeled probes originated from a bacterial chromosome (BAC) library.
Advantages
  • Dual colored probes for fast, sensitive, and specific detection
  • Work on metaphase spread, paraffin embedded, and frozen tissue
  • Identify gene amplification, loss, and translocation
  • High signal to noise ratio
  • Low cross reactivity
Specifications
Specifications
Store at 4°C in the dark.
Tube
Fluorescent In Situ Hybridization (Cell)
3q22.3/3q12.1
PIK3CB/CEN3q FISH Probe
Genomic DNA
Liquid
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
FITC, Texas Red
DAPI Counterstain (1500 ng/mL ) 250 uL
Approx. 310kb/500kb