Learn More
Phosphorous Acid, Honeywell Fluka™
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécification
Density | 1.651g/mL at 25°C |
Quantity | 100 g |
UN Number | 2834 |
Formula Weight | 82g/mol |
Percent Purity | 99% |
Chemical Name or Material | Phosphorous Acid |