Learn More
Fisherbrand™ Ion Selective Electrode
Product Code. 11719718 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Membrane:
Solid
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Membrane Unit Size
11719718 Solid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11719718
  • ONLINE EXCLUSIVE PRICE
    537.00€
    340.12€ / Pack of 1
    Save 196.88€
    36% Off
View all promotions
Estimated Shipment: 15-12-2026
Add to basket
Add to basket
Ion selective perchlorate electrode with a reproducibility greater than 2%.
  • Ion selective perchlorate electrode
  • Reproducibility better than 2%
  • Interchangeable membrane
  • S7 compliant per-head connection
  • All pH / mV meters or ionometers
Specifications
Description Ion Selective Electrode
pH Range 2.5 to 11
Membrane Solid