Learn More
BRANDTECH™ BRAND™ PD-Tip™ II Precision Dispenser Tips for Repeating Pipettes
Product Code. p-8806010 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
0.1 mL
0.5 mL
1 mL
1.25 mL
10 mL
12.5 mL
2.5 mL
25 mL
5 mL
50 mL
Unit Size:
Pack of 100
Pack of 25
Pack of 50
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
16368056 0.1 mL Pack of 100
16323166 0.5 mL Pack of 100
16333166 1 mL Pack of 100
16343166 1.25 mL Pack of 100
16353166 2.5 mL Pack of 100
16363166 5 mL Pack of 100
16373166 10 mL Pack of 100
16309635 12.5 mL Pack of 100
16383166 25 mL Pack of 50
16393166 50 mL Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 16368056 Supplier BRANDTECH™ BRAND™ Supplier No. 705700
Only null left
Add to basket
Edge
Add to basket
BRAND™ PD-Tip™ II fits most instruments using standard repeating tips.
  • Automatic size recognition by BRAND™ HandyStep™ Touch, HandyStep™ Touch S and HandyStep™ Electronic Pipettes
  • Adapter included for 25 mL and 50 mL sizes
  • Adapters are autoclavable at 121°C for 20 minutes
  • PD-Tips II are not autoclavable
These tips are suitable for the following dispensers:
  • BRAND Handystep™, BRAND Handystep™ S, HandyStep™ electronic, HandyStep™ touch, HandyStep™ touch S
  • Eppendorf Multipette™ 4780
  • Gilson Repetman™EDOS™ 5221
  • Rainin AutoRep™ E, Rainin AutoRep™ S, Rainin AutoRep™ M
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.1 mL
Filtered Non-filtered
Sterility Non-sterile
Color Natural
Autoclavable Not Autoclavable
Model PD-Tip II
Precision ≤5 to 3%
Accuracy ±≤8 to 1.6%
Certifications/Compliance Certificate of Performance
Clean Claims Virgin raw materials
Format Bulk, Bagged
Tip Style Positive Displacement
Packaging 100/Pk
Quantity 100
Product Type Precision Dispenser Tips
Disposable Single-use
For Use With (Equipment) Universal Fit Repeater Pipettor
Graduated Yes
Product Line PD-Tip II
Material Polypropylene, Polyethylene (Filter)
Graduations Printed