Learn More
Microspec™ Pasteur Pipette
Click to view available options
Volume (Metric) Dispense:
1 mL
3 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 500
Description
Bulk Pack of 500. Available as sterile or non-sterile.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Calibrated | No |
| Sterility | Sterile |
| Graduated | Yes |
| Quantity | 500 |
| Volume (Metric) Dispense | 1 mL |
| Color | Clear |
| Disposable | Single-use |