Learn More
Alla France™ Partial Immersion Precision Thermometer
Product Code. 11707479
Change view
Click to view available options
Temperature Range (Metric):
-20°C to +150°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) Unit Size
11707479 -20°C to +150°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11707479 Supplier Alla France™ Supplier No. 5120P150qp
  • ONLINE EXCLUSIVE PRICE
    16.96€
    9.70€ / Pack of 1
    Save 7.26€
    42% Off
In Stock
Add to basket
Edge
Add to basket
Use these thermometers for a variety of applications. Alla France™ Partial Immersion Precision Thermometers are manufactured using yellow Sodocalcique glass with a diameter of 6 to 7mm.
  • Mercury free : red liquid filling
  • With triangular ring
  • Expansion chamber supports temperatures higher than those of the thermometer
  • The graduation is printed and fused in the glass with a special process
  • Graduation resists to all chemical products except fluoridric acid and hot soda (to a lesser extent) because these 2 products attack the glass
  • Even if you can feel a relief when touching the graduation, the thermometer is not engraved
  • Available in partial or total immersion
  • Individually packed in recyclable sturdy plastic concertina box's, ideal for storing the instrument when not in use
  • Some references are also available in sealed blister by 6 pcs.
  • Systematically delivered with a Precision and Traceability Certificate
Specifications
Bottle Fill Red Liquid
Certifications/Compliance Precision and Traceability Certificate
Immersion Type Partial Immersion
Length (Metric) 305 mm
Material Soda Lime Glass
Product Type Precision Thermometer
Max. Temperature (Metric) 150°C
Min. Temperature (Metric) -20°C
Temperature Range (Metric) -20°C to +150°C
Accuracy ±1°C
Subdivision 1°C
Immersion Length 76 mm