Learn More
Cole-Parmer™ Parker Hannifin™ Stainless Steel Threaded Adapter
Use threaded fittings with our rigid tubing for your high-pressure applications
Brand: Cole-Parmer™ 31830-70
Additional Details : Weight : 0.80000kg
Description
- Threaded adapter.
- Stainless steel.
- 1/8in. female BSPP to 1/8in. male NPT.
- Threaded fittings are sturdy and designed for permanent installation.
- We offer threaded couplings and reducers, elbows and nipples, reducers and bushings, bushings, tees and plugs, and bulkhead and manifolds.
- Available in a variety of sizes.
Specifications
Threaded adapter, stainless steel, 1/8 in. female BSPP to 1/8 in. male NPT | |
BSPP(F) x NPT(M) | |
Stainless Steel |