Learn More
Fisherbrand™ Oval PTFE Stir Bar
Product Code. p-8007731 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
10 mm
15 mm
20 mm
25 mm
30 mm
35 mm
40 mm
50 mm
64 mm
70 mm
Diameter (Metric):
5 mm
6 mm
10 mm
12 mm
13 mm
16 mm
17 mm
20 mm
Unit Size:
Each
Pack of 10
Pack of 5
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
11898872 20 mm 10 mm Pack of 10
11517802 20 mm 10 mm Pack of 5
11858952 40 mm 13 mm Pack of 5
16285831 50 mm 20 mm Pack of 5
11828882 40 mm 20 mm Pack of 5
10603731 40 mm 20 mm Each
11338703 30 mm 16 mm Pack of 5
11808882 30 mm 16 mm Pack of 10
16295831 64 mm 20 mm Pack of 5
11507802 15 mm 6 mm Pack of 5
11828952 15 mm 6 mm Pack of 10
11868952 50 mm 17 mm Pack of 5
11838952 25 mm 10 mm Pack of 10
10445231 25 mm 12 mm Pack of 5
11844792 25 mm 12 mm Pack of 10
16245831 35 mm 16 mm Pack of 10
10609162 70 mm 20 mm Each
11888872 10 mm 5 mm Pack of 10
11597792 10 mm 5 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
Product Code. 11898872
Only null left
Add to basket
Edge
Aggiungi al carrello
Provides stable stirring action. Fisherbrand; Oval PTFE Stir Bar povides optimal turbulence for round-bottom flasks and containers.
  • Stable, general-purpose magnetic stir bar
  • Provides excellent turbulence for round-bottom flasks and containers
  • Autoclavable
Specifica
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Round bottom flasks
Diameter (Metric) 10 mm
Length (Metric) 20 mm
Quantity 5/Pk.
Shape Oval
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.