Learn More
Gilac™ Oval Proofing Basket
Oval Proofing Basket ensures that the dough is well ventilated; it can be staggered stacked and nested to save space.
24.33€ - 27.13€
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Dimensions (L x W x H) | Price | Quantity & Availability | ||||
17998801
|
Gilac™
G070180 |
0.5 kg | 21 x 15 x 9 cm |
24.33€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
17948881
|
Gilac™
G070190 |
0.75 kg | 25 x 15 x 9 cm |
26.31€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
17908501
|
Gilac™
G070210 |
1 kg | 26.5 x 17 x 9 cm |
27.13€
Each |
Estimated Shipment: 06-06-2024
Log in to see stock. |
||||
Description
- The cavity structure ensures that the dough is well ventilated.
- Can be staggered stacked and nested to save space.
- Comes with food contact certificates for optimum food safety and hygiene.
- Easy to clean and machine-washable.
- Light and extremely durable.
- Wide range of 10 product references to meet all needs (0.5 to 2 kg capacity).
- Liner (G072010) available as an option.
Specifications
Proofing Basket | |
Polypropylene | |
Oval |