Learn More
Brady™ Oval Polyethylene Tags
Tag a variety of assets with this tough polyethylene tag label material (B-145) that prints on both sides and folds to form an oval tag. Easily print anywhere with a Brady™ handheld label printer.
Brand: Brady™ 170851
Additional Details : Weight : 0.17700kg
Description
- Suitable for a wide range of applications including lockout and safety warning repair
- Designed to resist tearing and exposure to weather and extreme temperatures
- Simply print, fold, and easily attach using cable ties
Specifications
RoHS Compliant to 2005/618/EC MCV Amendment to RoHS Directive 2002/95/EC | |
Black on Gray | |
Die-cut | |
Labels | |
Asset Tracking, Component and Equipment Labeling, Data and Telecommunications Labeling, Facility Identification, General Identification, Inventory and Inspection Labeling, Safety Identification, Wire and Cable Labeling | |
65 Tags/Cartridge | |
0.023 in. | |
0.75 in. | |
3.1 cm |
Non-adhesive | |
Matte | |
Polyethylene | |
Non-sterile | |
BMP51, M511 | |
-40°F to +212°F | |
0.75 x 1.206 in. | |
1.206 in. |