Learn More
Thermo Scientific™ Orion™ Calcium Electrodes
Measure calcium in aqueous solutions simply, accurately and economically with these calcium electrodes.
Brand: Thermo Scientific™ 972001
265.28 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
Measure calcium in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Calcium Electrodes.
9720BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. To be used with module 972001.
A separate reference electrode is unnecessary with this electrode, making it convenient to use with small samples.
Related Products:
- Calibration standards: 922006 (0.1 M CaCl2); 923206 (100ppm)
- Required ISA: 932011
Specifications
Replacement Module for Combination Calcium Electrode 9720BNWP | |
Free-flowing Sure-Flow junction assures stable, drift-free potentials | |
ISE Meters | |
Combination Ion Selective Electrode |