Learn More
Thermo Scientific™ Orion™ Calcium Electrodes
Measure calcium in aqueous solutions simply, accurately and economically with these calcium electrodes.
Brand:  Thermo Scientific™ 932001
237.54 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10587211
  • 270.00€ / Each
Estimated Shipment: 03-05-2024
Description
Measure calcium in aqueous solutions simply, accurately and economically with Thermo Scientific™ Orion™ Calcium Electrodes.
9720BNWP is a combination electrode with a Sure-Flow™ junction. When conventional electrode junctions clog, the Sure-Flow junction can be opened and flushed clean simply by pressing the cap. To be used with module 972001.
A separate reference electrode is unnecessary with this electrode, making it convenient to use with small samples.
Related Products:
  • Calibration standards: 922006 (0.1 M CaCl2); 923206 (100ppm)
  • Required ISA: 932011
Specifications
Specifications
Replacement Module for Half-Cell Calcium Electrode 9320BN
Free-flowing Sure-Flow junction assures stable, drift-free potentials
ISE Meters
Combination Ion Selective Electrode