missing translation for 'onlineSavingsMsg'
Learn More

OPTIZYME™ PstI (6000U), Fisher BioReagents™

Brand:  Fisher Bioreagents BP8001-1

Additional Details : Weight : 0.01000kg

Product Code. 11994832

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

5'...C T G C A^G...3'
3'...G^A C G T C...5'

Conditions for 100% Activity:

  • 1X OPTIZYME Buffer 3: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2 , 100mM NaCl and 0.1mg/ml BSA.
  • Incubate at 37°C.
Supplied with: 10X OPTIZYME Buffer 3
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 50 -1 00%
  • Buffer 2: 50 - 100%
  • Buffer 3: 100%
  • Buffer 4: 50 - 100%
  • Buffer 5: 100%
Storage Buffer:
10mM Tris-HCl (pH 7.4 at 25°C), 200mM NaCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton X-100, 0.2mg/ml BSA and 50% (v/v) glycerol.
Compatible Ends: Alw21I, ApaI, BseSI, Eco24I, Mph1103I, PstI, SacI, SdaI.
Ligation and Recleavage:
More than 95% of DNA fragments can be ligated and recut after a 50-fold overdigestion with PstI.
Digestion of Agarose-embedded DNA:
A minimum of 5 units of PstI is required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Methylation Effects:
  • Dam: never overlaps - no effect.
  • Dcm: never overlaps - no effect.
  • CpG: never overlaps - no effect.
  • EcoKI: never overlaps - no effect.
  • EcoBI: never overlaps - no effect.

Specifications

Specifications

10 U/μL
37°C
>90% of DNA fragments can be ligated and re-cut after 10-fold over-digestion with Pst I
Keep container tightly closed
CTGCA.G
Liquid
>50% Glycerin, 25 to 50% Water, 1 to 2.5% Sodium Chloride
7.4
10mM Tris HCl (pH 7.4 at 25°C), 200mM NaCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton™ X-100, 0.2mg/mL BSA, 50% Glycerol
6000 U
PstI
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers