Learn More
Corning™ LSE™ Optional Head
Product Code. 11961981 Shop All Corning Products
Change view
Click to view available options
Holds:
12 x 1.5 to 2 mL Tubes
2 x 50 mL Tubes
4 x 15 mL Tubes
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds unitSize
11961981 12 x 1.5 to 2 mL Tubes Each
11981981 2 x 50 mL Tubes Each
11971981 4 x 15 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11961981 Supplier Corning™ Supplier No. 480105
Only null left
Add to basket
Add to basket
For use with various sizes of tubes.
This is an optional head for tubes, held horizontally, for use with Corning™ LSE™ Vortex Mixer.
TRUSTED_SUSTAINABILITY
Specifications
Holds 12 x 1.5 to 2 mL Tubes
Product Type Optional Head
For Use With (Equipment) Corning™ LSE™ Vortex Mixer