Learn More
Köttermann Open Tall Storage Cabinet
The open-front design of this Open Tall Storage Cabinet provides easy access to stored equipment and utensils.
Brand: Köttermann 301-00048
Additional Details : Weight : 60.55570kg
Description
- Tall, slender design expands storage with a small footprint
- Open front provides easy access to stored items
Specifications
None | |
Storage | |
No | |
60.6 kg | |
4 | |
Epoxy Coated | |
White |
None | |
No | |
500 x 600 x 1920 mm | |
None | |
Tall Storage | |
None | |
Steel |