Learn More
Zeiss™ Objective EC Plan-Neofluar
Enhanced Contrast “Plan-Neofluar” universal objectives with consequentliy minimized stray light. Use of glass types of low autofluorescence makes these objectives ideal for epi-fluorescence microscopy
Brand:  Zeiss™ 440350-9903-000
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11890893
  • 2883.20€ / Each
Estimated Shipment: 11-07-2024
Description
  • Visible increase in contrast important for all contrast methods: brightfield, darkfield, phase contrast, DIC, fluorescence, Pol
  • Feature high resolving power and brilliant, rich-contrast, completely flat images for observation and documentation
  • High transmission of wavelengths from the near UV range (>40% starting at 340nm)
  • Special-purpose members of the family are EC “Epiplan-Neofluar” objectives for reflected-light work
  • EC “Plan-Neofluar” Antiflex objective for reflection contrast
  • EC “Plan-Neofluar” Pol objectives for the requirements of polarizing microscopy
Specifications
Specifications
Objective EC Plan-Neofluar
0.71 mm
40X