Learn More
Motic™ Objective, Achromatic 20X/0.40
Designed with chromatic aberration correction for 2 colors. Motic™ Objective, Achromatic 20X/0.40 is spherically corrected for one color.
Brand: Motic™ 1101001701141
Additional Details : Weight : 0.01000kg
Description
- Achromatic.
- Chromatic aberration correction for 2 colors.
- Spherical correction for 1 color.
Specifications
Achromat Objective | |
NA: 0.40 | |
20X |