Learn More
Thermo Scientific™ Nunc™ Seals, Tapes, and Foils
Product Code. p-4525142
Change view
Click to view available options
Material:
Aluminum
Non-woven Rayon
PVC-coated Polypropylene
Plastic
Polyester
Polyethylene
Polyolefin
Polypropylene
Vinyl
For Use With (Application):
Breathable, Cell culture
Cell culture, sterile applications
Cell growth, Bacterial growth
Consistent seal
Crystallography and genotyping
Fluorescence, photosensitive samples
General purpose, ELISA
General purpose, sample detection
Low autofluorescence, ELISA
Luminescence, photosensitive samples
PCR
PCR, fluorescence assays, microscopy
PCR, long-term storage
PCR, qPCR, ELISA, Protein crystallography
PCR, storage in 384 well plates
PCR, storage in 96-well plates
Short-term storage, ELISA
Short-term storage, ELISA, minimizes evaporation and cross contamination
Ventilating and storing cell cultures
Certified RNase/DNase-free:
No
Yes
Unit Size:
Pack of 100
Pack of 1000
Pack of 2
Pack of 50
Pack of 800
19 product options available for selection
Product selection table with 19 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material For Use With (Application) Certified RNase/DNase-free Unit Size
10716943 Aluminum PCR, storage in 96-well plates Yes Pack of 100
10681864 Aluminum PCR, storage in 384 well plates Yes Pack of 100
10265411 Polypropylene PCR Yes Pack of 100
10194411 Polypropylene PCR, fluorescence assays, microscopy Yes Pack of 100
10633973 Polyethylene Low autofluorescence, ELISA No Pack of 100
10275452 Polyolefin PCR, qPCR, ELISA, Protein crystallography Yes Pack of 100
11371555 Vinyl Luminescence, photosensitive samples No Pack of 50
10170671 Polyester Cell culture, sterile applications No Pack of 800
11948231 Polyester General purpose, sample detection No Pack of 100
10160993 Vinyl Fluorescence, photosensitive samples No Pack of 50
10552615 Polyester General purpose, ELISA Yes Pack of 1000
10681344 Non-woven Rayon Breathable, Cell culture No Pack of 50
10082182 Non-woven Rayon Cell growth, Bacterial growth Yes Pack of 50
10777042 Plastic Consistent seal No Pack of 2
10574561 Aluminum PCR, long-term storage Yes Pack of 100
10333925 Polyester Short-term storage, ELISA No Pack of 100
10304315 Polyester Short-term storage, ELISA, minimizes evaporation and cross contamination No Pack of 100
10642674 Polypropylene Ventilating and storing cell cultures No Pack of 50
11524794 PVC-coated Polypropylene Crystallography and genotyping No Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 19 options available.
19 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Product Code. 10716943 Supplier Thermo Scientific™ Supplier No. 232698
Add to basket
Edge
Add to basket
Bulk Quote Request
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27970” to get your promotional price
Minimize evaporation and protect samples from contamination and spilling with these sealing tapes, ideal for all assay applications.
Minimize evaporation and protect samples from contamination and spilling with Thermo Scientific™ Nunc™ Sealing Tapes. These adhesive seals are ideal for all assay applications. Nunc Life Science products offer reliability and reproducible results, and are backed by industry-leading quality standards and certifications.
  • Adhesive seals effectively seal all microplate formats
  • Pressure sensitive adhesive is not tacky to touch, but forms a strong bond to the plate upon application of pressure
  • Thin, lightweight tapes seal onto the plate with a convenient, handheld applicator (available separately, Cat. No. 250050)
TRUSTED_SUSTAINABILITY
Specifications
Color Foil
Solvent Or Alcohol Resistant Life Science Reagent Resistant
Description AL SEAL TAPE FOR 96 WELL PLT
Material Aluminum
Sterility Non-sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Case 100
No. per Pack 100
Evaporation Barrier Malleable allows for visual verification of seal security, minimizes evaporation and cross contamination
For Use With (Application) PCR, storage in 96-well plates
Content And Storage General; Long term
Product Type Adhesive Seal
Product Line Nunc
Adhesive Acrylic
Certified RNase/DNase-free Yes
Pierceable Yes
Temperature Range (Metric) -80°C to +120°C
Well Shape All well shapes
Plate Compatibility All Microplates
Incubation Conditions General
Peelable No