missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Fluoromount Mounting Media
Applications: Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Brand: Novus Biologicals™ NB900-66726
Additional Details : Weight : 0.00970kg
Description
Applications: Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Frozen, Immunohistochemistry-ParaffinRecommended Storage
Store at room temperature
Specifications
Fluoromount Mounting Media |