Learn More
Hamilton™ Non-Filtered Low Retention Racked Pipette Tips
With surface walls completely free of occlusions and cavities so fluid retention is virtually eliminated.
Brand:  Hamilton™ 68010-22
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15375074
  • 82.35€ / Pack of 10
Estimated Shipment: 10-07-2024
to see stock.
Add to basket
Description
Description
  • Manufactured from chemically resistant virgin polypropylene for excellent clarity and smooth surfaces
  • Available pre-sterilized or non-sterile
  • 96 tips to a rack
  • Delivering outstanding accuracy and precision, guaranteed
Specifications
Specifications
1000 μL
Non-Filtered
Sterile
6 racks; 576 total tips