Learn More
R&L Enterprises™ Hoffman Clips
Product Code. 12823183 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
For Use With:
For use with 30mm tubing
For use with 40mm O.D. tubing
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
12823183 For use with 30mm tubing Pack of 10
12764396 For use with 40mm O.D. tubing Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12823183
Estimated Shipment: 30-12-2026
Add to basket
Add to basket
An excellent option for sealing flexible tubing in the laboratory. R&L Enterprises™ Hoffman Clips are made of nickel plated brass to prevent corrosion.
  • Either side or bottom opening
  • Available for a range of tube thicknesses
  • Threaded bar with thumbscrew
Specifications
Material Nickel-plated
For Use With For use with 30mm tubing
Packaging Quantity 10 Pack
Product Type Clip