Learn More
Bolle Safety Ness+ Platinum™ Safety Glasses
Product Code. p-7164939
Change view
Click to view available options
Lens Tint:
CSP (Comfort Sensitivity Perception)
Clear
Smoke
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint unitSize
16411881 Clear Each
16401881 Smoke Each
16491871 CSP (Comfort Sensitivity Perception) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16411881 Supplier Bolle Safety Supplier No. NESSPPSI
Only null left
Add to basket
Edge
Add to basket
Ness+ Platinum™ Safety Glasses featuring all the benefits of the NESS model with added comfort from co-injected temples.
  • Ultra-wraparound frame
  • PC/TPR co-injected temples for greater comfort
  • Panoramic field of vision
  • Scratch and fog resistant coating
  • Non-slip nose bridge
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE, EN166 FT
Lens Type Platinum™
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety Glasses
No. per Pack 1 Ea.
Weight (Metric) 28 g
Lens Material Polycarbonate