Learn More
Dr.Weigert™ Neodisher PolyKlar
Neodisher PolyKlar - Rinse aid liquid. Free of surfactants. Neutralizes alkaline residues during automated reprocessing. Designed for rinsing of animal cages.
Brand: Dr.Weigert™ 401930
134.10 EUR valid until 2024-05-31
Use promo code "21679" to get your promotional price.
Additional Details : Weight : 10.00000kg
Description
- Rinse aid Without surfactant
- neodisher PolyKlar is used in specially designed washers for animal cages and accessories
- Dosage of between 0.5-2 mL/L
- Acidic formulation
- Excellent material compatibility eg polysulfones, polycarbonates, polyphenylsulfones and polyetherimides
- Neutralizes alkaline residues
Specifications
Neodisher Polyklar | |
3.6 to 3.2 | |
Polycarboxylates, chloromethylisothiazolinones, methylisothiazolinones | |
1.0 g/cm3 (20°C) | |
Filtered |
Liquid | |
Non-sterile | |
0.5 to 2 mL/L | |
310 x 230 x 180 mm | |
10 L |
Safety and Handling
- neodisher PolyKlar
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : disodium metasilicate pentahydrate, sodium carbonate, troclosene sodium