Learn More
VACUUBRAND™ NBR Trapped O-Ring Centering Rings
10.35€ - 11.55€
Specifications
For Use With | For Small KF Flange |
---|---|
Material | NBR |
Quantity | 1 Each |
Type | Trapped O-ring Centering Rings |
Product Code | Brand | Diameter (Metric) Inside | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inside | Size | Price | Quantity & Availability | ||||
10203281
|
VACUUBRAND™
20660190 |
10/60 mm | 32 x 30.1 x 27.7 mm |
10.35€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10213281
|
VACUUBRAND™
20660191 |
20/25 mm | 42 x 40.1 x 36.7 mm |
11.55€
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
||||
Specifications
For Small KF Flange | |
NBR | |
1 Each | |
Trapped O-ring Centering Rings |