Learn More
Abeba™ Nature 8045 Shoes
Leather covered cork footbed is ESD dissipative.
Brand:  Abeba™ 8045/43
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 15622998
  • 87.16€ / 1 pair
Estimated Shipment: 03-06-2024
Description
  • CE EN ISO 20347:2012 OB A, FO SRC
  • Smooth leather
  • Black
  • Slip resistance special rubber outsole sole
  • Leather coated cork footbed
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Adjustable instep area
  • Buckle fastening
Specifications
Specifications
CE EN ISO 20347:2012 OB A, FO SRC
Cork
43
Rubber
Black
Leather
Slip-resistant
8045