Learn More
Saint Gobain Life Sciences Natural Rubber Folding Skirt Stoppers
Use these stoppers for extra protection of tube ends during transport, handling and other operations during which tubes require both internal and end protection. Saint Gobain Life Sciences™ Natural Rubber Folding Skirt Stoppers provide an excellent seal.
25.05€ - 45.10€
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
11704598
|
Saint Gobain Life Sciences
407018-20 |
11.5mmH/20mmH |
45.10€
Pack of 20 |
In Stock
Log in to see stock. |
|||||
11784588
|
Saint Gobain Life Sciences
407013-50 |
10mmH/20mmH |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11794588
|
Saint Gobain Life Sciences
407015-20 |
11mmH/20mm H |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11714598
|
Saint Gobain Life Sciences
407020-20 |
12.5mmH/19mmH |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11724598
|
Saint Gobain Life Sciences
40702510 |
18mmH/25mmH |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11774588
|
Saint Gobain Life Sciences
407010-50 |
8.5mmH/13mmH |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
11734598
|
Saint Gobain Life Sciences
407030-05 |
22mmH/27.5mmH |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
- Stoppers are flexible and provide a complete seal and good adhesion
- Grey-blue grade: made from natural rubber
- Temperature resistance from -35 to +70°C (120°C in moist heat)
Specifications
Natural Rubber |