Learn More
R&L Enterprises™ Semi-Micro Natural Gas Bunsen Burner
Product Code. 12852154 Shop All Fisherbrand Products
Change view
Click to view available options
Height (Metric):
100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) unitSize
12852154 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12852154
Only null left
Add to basket
Add to basket
An excellent solution for heating, sterilizing and combustion in the laboratory. R&L Enterprises™ Semi-Micro Natural Gas Bunsen Burners are constructed in brass and nickel plated.
  • Constructed using a heavy pressure diecast base painted in a chemically resistant blue acrylic finish
  • Tested for flame quality and leakage prior to packing
Specifications
Product Type Semi-micro Burner
Gas Type Methane, Natural Gas
Diameter (Metric) Outer Inlet 9 mm
Height (Metric) 100 mm
No. per Pack 1 Ea.
Weight (Metric) 0.22 kg