Learn More
IKA™ MT 40.100 Grinding Chamber
Product Code. 15153129 Shop All IKA Products
Change view
Click to view available options
Capacity (Metric):
40 mL
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15153129 40 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15153129 Supplier IKA™ Supplier No. 0020001173
  • ONLINE EXCLUSIVE PRICE
    922.00€
    830.00€ / Pack of 100
    Save 92.00€
    9% Off
In Stock
Add to basket
Edge
Add to basket
The MT 40.100 disposable grinding chamber with a stainless steel beater reduces soft, medium, hard and brittle materials with a Mohs hardness of up to 5 (manganese or apatite: 5 Mohs).
The chamber is made of transparent plastic so grinding tests can be observed at any time
TRUSTED_SUSTAINABILITY
Specifications
Product Type Grinding Chamber
Capacity (Metric) 40 mL