Learn More
Cytiva™ Whatman™ Microplus MP31 Nitrocellulose Filtration Membranes, with Grid
Whatman™ MicroPlus MP31 Filtration Membranes are recommended for most routine applications and are manufactured under strictly controlled conditions. Supplied as 135 mm x 240 mm sheets with imprinted grid pattern (white lines on black) to aid particle counting.
Brand:  Cytiva™ 10407766
Additional Details : Weight : 0.01100kg
Product Code. 12475252
  • 845.00€ / Each
Estimated Shipment: 06-09-2024
Description
  • Product Type: Filtration Membrane
  • Grade: MP31
  • Material: Nitrocellulose
  • Shape: Rectangular Sheets
  • Dimensions (Metric): 135 mm x 240 mm
  • Pore Size: 0.45μm
  • Quantity: 50 Pk.
Specifications
Specifications
Black
Nitrocellulose
Non-sterile
White
1Case
1 Ea.
Filtration Membrane
Single-use
0.45 μm
MP31
MP31
50 Pk.
Rectangular Sheets
135 x 240 mm
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 1