Learn More
MP Biomedicals™ TallPrep™ Lysing Matrix Y, 4.5 mL Tubes
Lysing Matrix Y contains 0.5 mm diameter yttria-stabilized zirconium oxide beads and is used to lyse yeast, fungi and algae.
173.84€ - 524.70€
Specifications
Bead Type | Yttria-stabilized Zirconium Oxide |
---|---|
Sample Type | Yeast, Fungi, Algae |
For Use With (Equipment) | FastPrep instruments or other bead beaters |
For Use With (Application) | Nucleic Acid Isolation and Purification, Homogenization |
Matrix | Lysing Matrix Y |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15212860
|
MP Biomedicals™
116977100 |
100 x 4.5 mL |
524.70€
Pack of 100 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
15202860
|
MP Biomedicals™
116977050 |
50 x 4.5 mL |
287.26€
Pack of 50 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
15722472
|
MP Biomedicals™
116977025 |
25 x 4.5 mL |
173.84€
Pack of 25 |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Lysing Matrix Y tubes can be used with FastPrep instruments or similar bead beaters. The yttria-stabilized zirconium oxide beads have a high density and hardness for cracking tough cell walls, yet a smooth surface for minimizing shearing.
- Tube size: 4.5 mL
- Samples types: Gram (+/-) bacteria, fungal tissue and spores, yeast, molds, filamentous algae and biofilms
- Matrix composition: 0.5 mm diameter yttria-stabilized zirconium oxide beads
Specifications
Yttria-stabilized Zirconium Oxide | |
FastPrep instruments or other bead beaters | |
Lysing Matrix Y |
Yeast, Fungi, Algae | |
Nucleic Acid Isolation and Purification, Homogenization |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.