Learn More
MP Biomedicals™ Hat Medium
Used for the selective growth of hybridoma cells for monoclonal antibody production.
Brand: MP Biomedicals™ 091680849
Additional Details : Weight : 2.26800kg
Specifications
0° to -20°C | |
HAT medium Supplement | |
Sterile | |
100 mL |