Learn More
MP Biomedicals™ Goat Serum: Normal Pool, Lyophilized, Sterile Filtered
Sterile, filtered, Lyophilized Serum
Brand: MP Biomedicals™ 08642921
146.48 EUR valid until 2024-12-31
Use promo code "23257" to get your promotional price.
Additional Details : Weight : 0.05000kg
Specifications
2° to 8°C | |
Goat Serum | |
Sterile | |
Immunocytochemistry and blocking |
Goat | |
50 mL | |
Filtration | |
Yes |