Learn More
R&D Systems™ Mouse Th2 Cell Multi-Color Flow Cytometry Kit
Designed for the flow cytometric analysis of mouse Th2 cells using four fluorochrome-conjugated antibodies
Brand:  R&D Systems™ FMC012
Additional Details : Weight : 0.09700kg
Product Code. 18712134
  • 947.06€ / 1 set
Estimated Shipment: 01-07-2024
Description
Single-step stainingIncludes conjugated antibodies and corresponding isotype controls: IL-4 R-CFS (Affinity-purified polyclonal antibody; goat IgG), STAT6-APC (Clone 253906; mouse IgG2A), IL-5-PE (Clone TRFK5; rat IgG1), CD4-PerCP (Clone GK1.5; rat IgG2B)
Specifications
Specifications
Store the unopened product at 2°C to 8°C. Do not use past expiration date.