Learn More
Molecular Sieve Uop Type 3A, Honeywell Fluka™
Beads
68.82€ - 195.09€
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15640580
|
Honeywell Fluka
02573-250G |
250 g |
68.82€
250g |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
15630580
|
Honeywell Fluka
02573-1KG |
1 kg |
195.09€
1kg |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured