Learn More
Molecular Sieve Type 4A, Technical, 1.6-2.5 mm Beads (8-12 Mesh), Fisher Chemical™
Click to view available options
Quantity:
500 g
2.5 kg
Specifications
Specifications
Color | Beige |
pH | 11 |
Packaging | Plastic powder jar |
Quantity | 500 g |
Physical Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'