Learn More
Hamilton™ Hitachi™ LaChrom™ Autosampler Syringes
Enables the automatic introduction of samples into the sample loop.
Brand: Hamilton™ 0160310
Additional Details : Weight : 0.50000kg
Description
- Provides improved reproducibility and speed
- For use with Hitachi™ LaChrom™ HPLC autosamplers
- Special termination
Specifications
Autoclavable | |
500 μL | |
Special |
Autosampler syringe | |
Hitachi LaChrom Autosamplers |