Learn More
VistaLab™ Technologies MLA™ Digital Pipette Seal Kit
Maintenance part
Brand: VistaLab™ Technologies 9094
Additional Details : Weight : 0.10000kg
Description
Seal kit for MLA™ Digital Pipettes - available for various volumesSpecifications
Seal kit | |
Non-sterile | |
MLA™ Digital 200 to 1000 μL | |
2 in. | |
1 in. |
Not Autoclavable | |
200 to 1000 μL | |
1 Ea. | |
1 in. |