Learn More
TFA Dostmann™ Mini Thermometer
Brand:  TFA Dostmann™ 30.2017.02
Additional Details : Weight : 0.03600kg
Product Code. 11879120
  • 35.00€ / Each
Estimated Shipment: 13-05-2024
Specifications
60°C
-10°C to +60°C
Digital LCD
-10°C
±1°C