Learn More
Fisherbrand™ Horizontal Wide Format Midi-Plus Gel Unit
Product Code. 11563382 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Gel Dimensions:
140L x 230 mm W
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gel Dimensions Unit Size
11563382 140L x 230 mm W Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11563382
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 23-10-2026
Add to basket
Add to basket
Includes
Gel unit, wide format, Midi-Plus, 1 x gel casting tray, 2 x 1.0mm 16 sample combs, buffer recirculation ports, power supply connectors and loading strips
For screening and analysis of wide range of samples.
  • For analytical and preparative studies of nucleic acids
  • Allows greater number of samples to be run on one gel
  • Features four comb positions for fast separation of samples
  • Removable gel casting tray
Specifications
Type Horizontal Wide Format Midi-Plus Gel Unit
Gel Dimensions 140L x 230 mm W
Includes Gel unit, wide format, Midi-Plus, 1 x gel casting tray, 2 x 1.0mm 16 sample combs, buffer recirculation ports, power supply connectors and loading strips
Sample Size 200
Volume (Metric) Buffer 800 mL